O-tert-Butyl-L-tyrosine 99%

Supplier: Thermo Scientific

Synonyms: H-Tyr(tBu)-OH

B22575.03 B22575.06
B22575.03EA 91.5 EUR
B22575.03 B22575.06
O-tert-Butyl-L-tyrosine 99%
O-tert-Butyl-L-tyrosine

Purity: 99% CAS No.: 18822-59-8 Molecular Formula: C13H19NO3 Formula Weight: 237.3 Melting Point: ca 205° dec. MDL No.: MFCD00037202

Formula: C₁₃H₁₉NO₃
MW: 237,30 g/mol
Melting Pt: ∼ 205 °C (dec.)
Storage Temperature: Ambient
MDL Number: MFCD00037202
CAS Number: 18822-59-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR