3-(5-Methyl-2-furyl)butyraldehyde 99%

Supplier: Thermo Fisher Scientific
Warning

B22671.06 B22671.14
B22671.06EA 33.6 EUR
B22671.06 B22671.14
3-(5-Methyl-2-furyl)butyraldehyde 99%
3-(5-Methyl-2-furyl)butyraldehyde

Purity: 99% CAS No.: 31704-80-0 Molecular Formula: C9H12O2 Formula Weight: 152.19 Flash Point: 58°(136°F) Boiling Point: 93-94°/14mm UN No.: UN1989 MDL No.: MFCD00209519

Formula: C₉H₁₂O₂
MW: 152.19 g/mol
Boiling Pt: 93…94 °C (14 mmHg)
Density: 1.006
Flash Pt: 58 °C (136 °F)
Storage Temperature: Ambient
MDL Number: MFCD00209519
CAS Number: 31704-80-0
EINECS: 250-771-4
ADR: 3 III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR