2,2,3,3-Tetrafluoropropyl methacrylate 97% stabilised
Supplier: Thermo Scientific
|
Warning
|
Synonyms:
Methacrylic Acid 2,2,3,3-Tetrafluoropropyl Ester
Stabilisation:
Stabilised with 4-methoxyphenol (MEHQ) 50 ppm
B22682.06
B22682.06EA
35.1
EUR
B22682.06
2,2,3,3-Tetrafluoropropyl methacrylate 97% stabilised
2,2,3,3-Tetrafluoropropyl methacrylate
Stab. with 50ppm 4-methoxyphenol
Purity: 97%
CAS No.: 45102-52-1
Molecular Formula: C7H8F4O2
Formula Weight: 200.13
Flash Point: 50°(122°F)
Refractive Index: 1.3750
Boiling Point: 146-149°
Beilstein Reference: 3588831
UN No.: UN3272
MDL No.: MFCD00042381
Formula:
C₇H₈F₄O₂ MW: 200,13 g/mol Boiling Pt: 146…149 °C Density: 1,254 Flash Pt: 50 °C (122 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00042381 CAS Number: 45102-52-1 EINECS: 256-189-7 ADR: 3,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...