Sodium hexanitritoiridate(III) 99%

Supplier: Thermo Scientific
Warning

B23002.02 B23002.88
B23002.02EA 0 EUR
B23002.02 B23002.88
Sodium hexanitritoiridate(III) 99%
Sodium hexanitritoiridate(III)

Purity: 99% CAS No.: 94022-51-2 Molecular Formula: IrN6Na3O12 Formula Weight: 537.22 MDL No.: MFCD00064621

Formula: IrN₆Na₃O₁₂
MW: 537,22 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00064621
CAS Number: 94022-51-2
EINECS: 301-548-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR