(+)-Griseofulvin 97%

Supplier: Thermo Fisher Scientific
Danger

Synonyms: Griseofulvin

B23521.06 B23521.14 B23521.22
B23521.06EA 46.2 EUR
B23521.06 B23521.14 B23521.22
(+)-Griseofulvin 97%
(+)-Griseofulvin

Purity: 97% CAS No.: 126-07-8 Molecular Formula: C17H17ClO6 Formula Weight: 352.77 Storage Temperature: 2°C to 8°C Odour: Odourless Melting Point: 218-220° Beilstein Reference: 95226 MDL No.: MFCD00082343

Formula: C₁₇H₁₇ClO₆
MW: 352.77 g/mol
Melting Pt: 218…220 °C
MDL Number: MFCD00082343
CAS Number: 126-07-8
EINECS: 204-767-4

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR