Ethyl-4-phenylpyrrole-3-carboxylate 97%

Supplier: Thermo Scientific
Warning

Synonyms: Ethyl 4-phenyl-1H-pyrrole-3-carboxylate

B24877.03 B24877.06 B24877.14
B24877.03EA 53.3 EUR
B24877.03 B24877.06 B24877.14
Ethyl-4-phenylpyrrole-3-carboxylate 97%
Ethyl-4-phenylpyrrole-3-carboxylate

Purity: 97% CAS No.: 64276-62-6 Molecular Formula: C13H13NO2 Formula Weight: 215.25 Melting Point: 126-128° MDL No.: MFCD03931521

Formula: C₁₃H₁₃NO₂
MW: 215,25 g/mol
Melting Pt: 126…128 °C
Storage Temperature: Ambient
MDL Number: MFCD03931521
CAS Number: 64276-62-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR