Methyl-5-chloro-5-oxovalerate 97%
Supplier: Thermo Scientific
|
Danger
|
Synonyms:
Methyl glutaryl chloride, Methyl 4-chloroformylbutyrate
B25141.09
B25141.18
B25141.30
B25141.09EA
72.3
EUR
B25141.09
B25141.18
B25141.30
Methyl-5-chloro-5-oxovalerate 97%
Methyl-5-chloro-5-oxovalerate
Purity: 97%
CAS No.: 1501-26-4
Molecular Formula: C6H9ClO3
Formula Weight: 164.59
Storage Temperature: 2°C to 8°C
Refractive Index: 1.4470
Boiling Point: 108-110°/15mm
UN No.: UN3265
MDL No.: MFCD00000756
Formula:
C₆H₉ClO₃ MW: 164,59 g/mol Boiling Pt: 108…110 °C (15 mmHg) |
MDL Number:
MFCD00000756 CAS Number: 1501-26-4 EINECS: 216-115-6 ADR: 8,II |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...