Methyl phenyl carbonate 97%
Supplier: Thermo Scientific
|
Warning
|
B25232.03
B25232.06
B25232.14
B25232.03EA
37
EUR
B25232.03
B25232.06
B25232.14
Methyl phenyl carbonate 97%
Methyl phenyl carbonate
Purity: 97%
CAS No.: 13509-27-8
Molecular Formula: C8H8O3
Formula Weight: 152.15
Refractive Index: 1.521
Boiling Point: 210-213°
MDL No.: MFCD04039782
Formula:
C₈H₈O₃ MW: 152,15 g/mol Boiling Pt: 210…213 °C Storage Temperature: Ambient |
MDL Number:
MFCD04039782 CAS Number: 13509-27-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...