2,3-Dibromo-1,4-butanediol ≥98%
Supplier: Thermo Scientific
B25260.14
B25260.22
B25260.14EA
0
EUR
B25260.14
B25260.22
2,3-Dibromo-1,4-butanediol ≥98%
2,3-Dibromo-1,4-butanediol
Purity: 98%
CAS No.: 20163-90-0
Molecular Formula: C4H8Br2O2
Formula Weight: 247.93
Melting Point: 83-87°
Boiling Point: 148-150°/1.5mm
MDL No.: MFCD00011549
Formula:
C₄H₈Br₂O₂ Melting Pt: 83…87 °C |
MDL Number:
MFCD00011549 CAS Number: 20163-90-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...