3-(Ethoxymethylene)pentane-2,4-dione ≥98%
Supplier: Thermo Scientific
B25445.14
B25445.06
B25445.14EA
205
EUR
B25445.14
B25445.06
3-(Ethoxymethylene)pentane-2,4-dione ≥98%
3-(Ethoxymethylene)pentane-2,4-dione
Purity: 98+%
CAS No.: 33884-41-2
Molecular Formula: C8H12O3
Formula Weight: 156.18
Refractive Index: 1.484
Boiling Point: 140-142°/15mm
MDL No.: MFCD07368652
Formula:
C₈H₁₂O₃ Boiling Pt: 140…142 °C (15 mmHg) Storage Temperature: Ambient |
MDL Number:
MFCD07368652 CAS Number: 33884-41-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...