1-Methyl-2-formylbenzimidazole ≥98%

Supplier: Thermo Scientific
Warning

Synonyms: 1-Methyl-1H-benzo[d]imidazole-2-carbaldehyde

B25601.03 B25601.06
B25601.03EA 118 EUR
B25601.03 B25601.06
1-Methyl-2-formylbenzimidazole ≥98%
1-Methyl-2-formylbenzimidazole

Purity: 98+% CAS No.: 3012-80-4 Molecular Formula: C9H8N2O Formula Weight: 160.17 Melting Point: 120-122° MDL No.: MFCD00142655

Formula: C₉H₈N₂O
MW: 160,176 g/mol
Melting Pt: 120…122 °C
Storage Temperature: Ambient
MDL Number: MFCD00142655
CAS Number: 3012-80-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR