Ethyl-2-pentynyl ether 97%

Supplier: Thermo Scientific
Warning

H25960.14 H25960.06
H25960.14EA 0 EUR
H25960.14 H25960.06
Ethyl-2-pentynyl ether 97%
Ethyl-2-pentynyl ether

Purity: 97% CAS No.: 20635-10-3 Molecular Formula: C7H12O Formula Weight: 112.17 Flash Point: 30°(86°F) Boiling Point: 63-65°/20mm UN No.: UN3271 MDL No.: MFCD00048702

Formula: C₇H₁₂O
MW: 112,17 g/mol
Boiling Pt: 63…65 °C (20 mmHg)
Density: 0,864
Flash Pt: 30 °C (86 °F)
Storage Temperature: Ambient
MDL Number: MFCD00048702
CAS Number: 20635-10-3
ADR: 3,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR