2-Amino-4-chloro-6-isopropylpyrimidine 97%
|
Warning
|
Synonyms:
4-Chloro-6-isopropylpyrimidin-2-amine
H25997.MD
H25997.03
H25997.MDEA
93
EUR
H25997.MD
H25997.03
2-Amino-4-chloro-6-isopropylpyrimidine 97%
2-Amino-4-chloro-6-isopropylpyrimidine
Purity: 97%
CAS No.: 73576-33-7
Molecular Formula: C7H10ClN3
Formula Weight: 171.63
Melting Point: 98-101°
MDL No.: MFCD03840198
Formula:
C₇H₁₀ClN₃ MW: 171,63 g/mol Melting Pt: 98…101 °C Storage Temperature: Ambient |
MDL Number:
MFCD03840198 CAS Number: 73576-33-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...