α-2-Dimethylstyrene 99%
Supplier: Thermo Scientific
|
Warning
|
H26073.06
H26073.03
H26073.06EA
227
EUR
H26073.06
H26073.03
α-2-Dimethylstyrene 99%
alpha-2-Dimethylstyrene
Purity: 99%
CAS No.: 26444-18-8
Molecular Formula: C10H12
Formula Weight: 132.21
Storage Temperature: 2°C to 8°C
Flash Point: 43°(110°F)
Refractive Index: 1.515
Boiling Point: 53-55°/11mm
UN No.: UN3295
MDL No.: MFCD00049092
Formula:
C₁₀H₁₂ MW: 132,21 g/mol Boiling Pt: 53…55 °C (11 mmHg) Density: 0,894 Flash Pt: 43 °C (110 °F) |
MDL Number:
MFCD00049092 CAS Number: 26444-18-8 ADR: 3,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...