2-Chloro-5-hydroxybenzoic acid 99%

Supplier: Thermo Scientific
Warning

H26112.MD H26112.03
H26112.MDEA 0 EUR
H26112.MD H26112.03
2-Chloro-5-hydroxybenzoic acid 99%
2-Chloro-5-hydroxybenzoic acid

Purity: 99% CAS No.: 56961-30-9 Molecular Formula: C7H5ClO3 Formula Weight: 172.57 Melting Point: 178-180° MDL No.: MFCD04038818

Formula: C₇H₅ClO₃
MW: 172,57 g/mol
Melting Pt: 178…180 °C
Storage Temperature: Ambient
MDL Number: MFCD04038818
CAS Number: 56961-30-9
EINECS: None

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR