Methyl-3-amino-4-cyano-5-(methylthio)thiophene-2-carboxylate 97%

Supplier: Thermo Fisher Scientific
Danger

Synonyms: 3-Amino-4-cyano-5-(methylthio)thiophene-2-carboxylic acid methyl ester

H26233.03 H26233.09
H26233.03EA 0 EUR
H26233.03 H26233.09
Methyl-3-amino-4-cyano-5-(methylthio)thiophene-2-carboxylate 97%
Methyl-3-amino-4-cyano-5-(methylthio)thiophene-2-carboxylate

Purity: 97% CAS No.: 129332-45-2 Molecular Formula: C8H8N2O2S2 Formula Weight: 228.28 Melting Point: 205-208° UN No.: UN3439 MDL No.: MFCD00052752

Formula: C₉H₁₀N₂O₂S₂
Melting Pt: 205…208 °C
Storage Temperature: Ambient
MDL Number: MFCD00085050
CAS Number: 129332-45-2
UN: 3439
ADR: 6.1,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR