Dimethyl cyclohexylmalonate 95%
Supplier: Thermo Fisher Scientific
H26289.03
H26289.06
H26289.03EA
72.7
EUR
H26289.03
H26289.06
Dimethyl cyclohexylmalonate 95%
Dimethyl cyclohexylmalonate
Purity: 95%
CAS No.: 49769-76-8
Molecular Formula: C11H18O4
Formula Weight: 214.26
Refractive Index: 1.4565
MDL No.: MFCD05666206
Formula:
C₁₁H₁₈O₄ MW: 214.26 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD05666206 CAS Number: 49769-76-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...