N-Boc-D-prolinal 97%

Supplier: Thermo Scientific
Warning

Synonyms: 1-Pyrrolidinecarboxylic acid, 2-formyl-, 1,1-dimethylethyl ester, (2R)-, tert-Butyl (R)-2-Formylpyrrolidine-1-carboxylate, (R)-N-BOC-Prolinal, (R)-2-Formylpyrrolidine-1-carboxylic acid tert-Butyl ester, N-Boc-D-Prolinal, N-Boc-D-prolinal, N-(tert-Butoxycarbonyl)-D-prolinal, (R)-tert-butyl 2-formylpyrrolidine-1-carboxylate

H26294.06 H26294.03
H26294.06EA 689 EUR
H26294.06 H26294.03
N-Boc-D-prolinal 97%
N-Boc-D-prolinal

Purity: 97% CAS No.: 73365-02-3 Molecular Formula: C10H17NO3 Formula Weight: 199.25 Storage Temperature: -30°C to -10°C Flash Point: 110°(230°F) Refractive Index: 1.4618 Boiling Point: 228° Optical Rotation: +88° (c=1 in chloroform) MDL No.: MFCD01321389

Formula: C₁₀H₁₇NO₃
MW: 199,25 g/mol
Boiling Pt: 228 °C
Density: 1,06
Flash Pt: >110 °C (230 °F)
Storage Temperature: Freezer
MDL Number: MFCD01321389
CAS Number: 73365-02-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR