p-Quinquephenyl 98%

Supplier: Thermo Fisher Scientific

H26308.MD H26308.03
H26308.MDEA 0 EUR
H26308.MD H26308.03
p-Quinquephenyl 98%
p-Quinquephenyl

Purity: 98% CAS No.: 3073-05-0 Molecular Formula: C30H22 Formula Weight: 382.50 MDL No.: MFCD00059012

Formula: C₃₀H₂₂
MW: 382.51 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00059012
CAS Number: 3073-05-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR