Naphthalene-2-thiocarboxamide 97%

Supplier: Thermo Scientific
Warning

H26324.MD H26324.03
H26324.MDEA 47.8 EUR
H26324.MD H26324.03
Naphthalene-2-thiocarboxamide 97%
Naphthalene-2-thiocarboxamide

Purity: 97% CAS No.: 6967-89-1 Molecular Formula: C11H9NS Formula Weight: 187.26 Melting Point: 150-152° UN No.: UN2811 MDL No.: MFCD01571356

Formula: C₁₁H₉NS
MW: 187,26 g/mol
Melting Pt: 150…152 °C
Storage Temperature: Ambient
MDL Number: MFCD01571356
CAS Number: 6967-89-1
ADR: 6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR