2-Naphthalen-2-ylpiperazine 95%

Supplier: Thermo Scientific
Danger

H26453.MD H26453.03
H26453.MDEA 0 EUR
H26453.MD H26453.03
2-Naphthalen-2-ylpiperazine 95%
2-Naphthalen-2-ylpiperazine

Purity: 95% CAS No.: 904816-32-6 Molecular Formula: C14H16N2 Formula Weight: 212.30 Melting Point: 99-104° UN No.: UN3259 MDL No.: MFCD05864679

Formula: C₁₄H₁₆N₂
MW: 212,30 g/mol
Melting Pt: 99…104 °C
Storage Temperature: Ambient
MDL Number: MFCD05864679
CAS Number: 904816-32-6
ADR: 8,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR