Dimethyl tert-butylmalonate 95%

Supplier: Thermo Scientific

H26532.03 H26532.06
H26532.03EA 57.3 EUR
H26532.03 H26532.06
Dimethyl tert-butylmalonate 95%
Dimethyl tert-butylmalonate

Purity: 95% CAS No.: 39520-25-7 Molecular Formula: C9H16O4 Formula Weight: 188.22 Refractive Index: 1.4280 MDL No.: MFCD08436110

Formula: C₉H₁₆O₄
MW: 188,22 g/mol
Storage Temperature: Ambient
MDL Number: MFCD08436110
CAS Number: 39520-25-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR