2-Acetylquinoxaline 97%
Supplier: Thermo Fisher Scientific
|
Warning
|
H26680.MD
H26680.03
H26680.MDEA
0
EUR
H26680.MD
H26680.03
2-Acetylquinoxaline 97%
2-Acetylquinoxaline
Purity: 97%
CAS No.: 25594-62-1
Molecular Formula: C10H8N2O
Formula Weight: 172.19
Melting Point: 76-79°
MDL No.: MFCD07808559
Formula:
C₁₀H₈N₂O Melting Pt: 76…79 °C Storage Temperature: Ambient |
MDL Number:
MFCD07808559 CAS Number: 25594-62-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...