1-Butenyl ethyl ether cis- and trans-mixture 98%

Supplier: Thermo Scientific
Danger

H26720.06 H26720.14 H26720.22
H26720.06EA 0 EUR
H26720.06 H26720.14 H26720.22
1-Butenyl ethyl ether cis- and trans-mixture 98%
1-Butenyl ethyl ether

Purity: 98% CAS No.: 929-05-5 Molecular Formula: C6H12O Formula Weight: 100.16 Flash Point: 6°(43°F) Refractive Index: 1.4060 Boiling Point: 94-95° UN No.: UN3271 MDL No.: MFCD03427069

Formula: C₆H₁₂O
MW: 100,16 g/mol
Boiling Pt: 94…95 °C
Density: 0,779
Flash Pt: 6 °C (43 °F)
Storage Temperature: Ambient
MDL Number: MFCD03427069
CAS Number: 929-05-5
UN: 3271
ADR: 3,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR