2-Thiophen-2-yl-piperazine 96%

Supplier: Thermo Fisher Scientific
Warning

H26733.MD H26733.03
H26733.MDEA 0 EUR
H26733.MD H26733.03
2-Thiophen-2-yl-piperazine 96%
2-Thiophen-2-yl-piperazine

Purity: 96% CAS No.: 85803-49-2 Molecular Formula: C8H12N2S Formula Weight: 168.26 Melting Point: 82-84° MDL No.: MFCD06245415

Formula: C₈H₁₂N₂S
Melting Pt: 82…84 °C
Storage Temperature: Ambient
MDL Number: MFCD06245415
CAS Number: 85803-49-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR