6-Methyl-2-pyridinemethanol 98%
Supplier: Thermo Fisher Scientific
|
Warning
|
H26794.06
H26794.14
H26794.06EA
105
EUR
H26794.06
H26794.14
6-Methyl-2-pyridinemethanol 98%
6-Methyl-2-pyridinemethanol
Purity: 98%
CAS No.: 1122-71-0
Molecular Formula: C7H9NO
Formula Weight: 123.15
Flash Point: 109°(228°F)
Melting Point: 32-34°
Boiling Point: 105-108°/12mm
MDL No.: MFCD00023520
Formula:
C₇H₉NO Boiling Pt: 105…108 °C (12 mmHg) Melting Pt: 32…34 °C Flash Pt: 109 °C (228 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00023520 CAS Number: 1122-71-0 EINECS: 214-358-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...