D-Valine methyl ester hydrochloride 99%
Supplier: Thermo Scientific
Synonyms:
H-D-Val-OMe.HCl, H-D-Val-OMe·HCl
H26883.03
H26883.06
H26883.14
H26883.03EA
31.2
EUR
H26883.03
H26883.06
H26883.14
D-Valine methyl ester hydrochloride 99%
D-Valine methyl ester hydrochloride
Purity: 99%
CAS No.: 7146-15-8
Molecular Formula: C6H13NO2.HCl
Formula Weight: 167.64
Storage Temperature: 2°C to 8°C
Melting Point: ca 170° dec.
Beilstein Reference: 3912091
MDL No.: MFCD00237309
Formula:
C₆H₁₃NO₂ MW: 167,633 g/mol Melting Pt: ∼ 170 °C (dec.) |
MDL Number:
MFCD00237309 CAS Number: 7146-15-8 EINECS: 230-454-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...