1-Phenylsulphonyl-1H-pyrazole-4-boronic acid pinacol ester 95%
Supplier: Thermo Scientific
Synonyms:
1-(Phenylsulphonyl)-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-1H-pyrazole
H26954.MD
H26954.03
H26954.MDEA
147
EUR
H26954.MD
H26954.03
1-Phenylsulphonyl-1H-pyrazole-4-boronic acid pinacol ester 95%
1-Phenylsulphonyl-1H-pyrazole-4-boronic acid pinacol ester
Purity: 95%
CAS No.: 1073372-04-9
Molecular Formula: C15H19BN2O4S
Formula Weight: 334.20
Melting Point: 125-130°
MDL No.: MFCD09037504
Formula:
C₁₅H₁₉BN₂O₄S MW: 334,20 g/mol Melting Pt: 125…130 °C Storage Temperature: Ambient |
MDL Number:
MFCD09037504 CAS Number: 1073372-04-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...