2-Methanesulphonyl-4,6-dimethoxypyrimidine 96%
Supplier: Thermo Fisher Scientific
|
Warning
|
Synonyms:
4,6-Dimethoxy-2-(methylsulphonyl)pyrimidine
H27003.03
H27003.06
H27003.14
H27003.03EA
0
EUR
H27003.03
H27003.06
H27003.14
2-Methanesulphonyl-4,6-dimethoxypyrimidine 96%
2-Methanesulphonyl-4,6-dimethoxypyrimidine
Purity: 96%
CAS No.: 113583-35-0
Molecular Formula: C7H10N2O4S
Formula Weight: 218.23
Melting Point: 129-133°
MDL No.: MFCD00672151
Formula:
C₇H₁₀N₂O₄S Melting Pt: 129…133 °C Storage Temperature: Ambient |
MDL Number:
MFCD00672151 CAS Number: 113583-35-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...