4-Methyl-2H-1,4-benzoxazin-3(4H)-one 98%

Supplier: Thermo Scientific
Warning

Synonyms: 4-Methyl-2H-1,4-Benzoxazin-3-one

H27196.14 H27196.06
H27196.14EA 107 EUR
H27196.14 H27196.06
4-Methyl-2H-1,4-benzoxazin-3(4H)-one 98%
4-Methyl-2H-1,4-benzoxazin-3(4H)-one

Purity: 98% CAS No.: 21744-84-3 Molecular Formula: C9H9NO2 Formula Weight: 163.18 Melting Point: 58-60° MDL No.: MFCD00601365

Formula: C₉H₉NO₂
MW: 163,18 g/mol
Melting Pt: 58…60 °C
Storage Temperature: Ambient
MDL Number: MFCD00601365
CAS Number: 21744-84-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR