2-Mercapto-5-n-propylpyrimidine 98%
Supplier: Thermo Fisher Scientific
|
Warning
|
H27239.06
H27239.03
H27239.14
H27239.06EA
136
EUR
H27239.06
H27239.03
H27239.14
2-Mercapto-5-n-propylpyrimidine 98%
2-Mercapto-5-n-propylpyrimidine
Purity: 98%
CAS No.: 52767-84-7
Molecular Formula: C7H10N2S
Formula Weight: 154.24
Melting Point: 207-211°
MDL No.: MFCD07777095
Formula:
C₇H₁₀N₂S Melting Pt: 207…211 °C Storage Temperature: Ambient |
MDL Number:
MFCD07777095 CAS Number: 52767-84-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...