Dimethyl isopropenyl phosphate 95%
Supplier: Thermo Fisher Scientific
H27353.22
H27353.14
H27353.22EA
0
EUR
H27353.22
H27353.14
Dimethyl isopropenyl phosphate 95%
Dimethyl isopropenyl phosphate
Purity: 95%
CAS No.: 4185-82-4
Molecular Formula: C5H11O4P
Formula Weight: 166.11
Refractive Index: 1.4200
Boiling Point: 120°/7.5mm
MDL No.: MFCD09953485
Formula:
C₅H₁₁O₄P Boiling Pt: 120 °C (7,5 mmHg) Storage Temperature: Ambient |
MDL Number:
MFCD09953485 CAS Number: 4185-82-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...