tert-Butyldichlorophosphine 98%

Supplier: Thermo Scientific
Danger

Synonyms: t-Butyldichlorophosphine

H27360.03 H27360.09 H27360.18
H27360.03EA 40.4 EUR
H27360.03 H27360.09 H27360.18
tert-Butyldichlorophosphine 98%
tert-Butyldichlorophosphine

Purity: 98% CAS No.: 25979-07-1 Molecular Formula: C4H9Cl2P Formula Weight: 159.00 Melting Point: 44-49° UN No.: UN3261 MDL No.: MFCD00013615

Formula: C₄H₉Cl₂P
MW: 158,99 g/mol
Melting Pt: 44…49 °C
Storage Temperature: Ambient
MDL Number: MFCD00013615
CAS Number: 25979-07-1
EINECS: 247-386-9
ADR: 8,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR