2-(4-Ethylphenyl)-5-n-propylpyrimidine ≥99%
Supplier: Thermo Fisher Scientific
H27361.03
H27361.09
H27361.03EA
0
EUR
H27361.03
H27361.09
2-(4-Ethylphenyl)-5-n-propylpyrimidine ≥99%
2-(4-Ethylphenyl)-5-n-propylpyrimidine
Purity: 99+%
CAS No.: 98495-11-5
Molecular Formula: C15H18N2
Formula Weight: 226.32
Melting Point: 41-43°
MDL No.: MFCD00799380
Formula:
C₁₅H₁₈N₂ Melting Pt: 41…43 °C Storage Temperature: Ambient |
MDL Number:
MFCD00799380 CAS Number: 98495-11-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...