6-Bromo-2,4-dimethylpyridine-3-boronic acid 95%

Supplier: Thermo Fisher Scientific
Warning

H27460.MD H27460.03
H27460.MDEA 180 EUR
H27460.MD H27460.03
6-Bromo-2,4-dimethylpyridine-3-boronic acid 95%
6-Bromo-2,4-dimethylpyridine-3-boronic acid
Formula: C₇H₉BBrNO₂
Melting Pt: ∼ 145 °C (dec.)
Storage Temperature: Ambient
MDL Number: MFCD09037491
CAS Number: 1072944-23-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR