1-[(4-Ethoxyphenyl)ethynyl]-4-n-pentylbenzene ≥99%

Supplier: Thermo Scientific

Synonyms: 1-Ethoxy-4-((4-pentylphenyl)ethynyl)benzene

H27476.09 H27476.03
H27476.09EA 0 EUR
H27476.09 H27476.03
1-[(4-Ethoxyphenyl)ethynyl]-4-n-pentylbenzene ≥99%
1-[(4-Ethoxyphenyl)ethynyl]-4-n-pentylbenzene

Purity: 99+% CAS No.: 95480-29-8 Molecular Formula: C21H24O Formula Weight: 292.42 Melting Point: 62°(N) Boiling Point: 178-180°/0.5mm MDL No.: MFCD01218020

Formula: C₂₁H₂₄O
MW: 292,422 g/mol
Boiling Pt: 178…180 °C (0,5 mmHg)
Melting Pt: 62 °C(N)
Storage Temperature: Ambient
MDL Number: MFCD01218020
CAS Number: 95480-29-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR