1-Cyclopropyl-4-piperidinone 98%
Supplier: Thermo Fisher Scientific
Synonyms:
1-Cyclopropylpiperidin-4-one
H27495.06
H27495.14
H27495.03
H27495.06EA
185
EUR
H27495.06
H27495.14
H27495.03
1-Cyclopropyl-4-piperidinone 98%
1-Cyclopropyl-4-piperidinone
Purity: 98%
CAS No.: 62813-01-8
Molecular Formula: C8H13NO
Formula Weight: 139.20
MDL No.: MFCD07374375
Formula:
C₈H₁₃NO Storage Temperature: Ambient |
MDL Number:
MFCD07374375 CAS Number: 62813-01-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...