1,1'-Dibutylzirconocene dichloride ≥98%

Supplier: Thermo Scientific
Warning

Synonyms: Bis(n-butylcyclopentadienyl)zirconium dichloride, Bis(butylcyclopentadienyl)zirconium(IV) Dichloride

H27576.03 H27576.06
H27576.03EA 123 EUR
H27576.03 H27576.06
1,1'-Dibutylzirconocene dichloride ≥98%
1,1'-Dibutylzirconocene dichloride

Purity: 98+% CAS No.: 73364-10-0 Molecular Formula: C18H26Cl2Zr Formula Weight: 404.53 Melting Point: 97-99° MDL No.: MFCD00672105

Formula: C₁₈H₂₆Cl₂Zr
MW: 404,53 g/mol
Melting Pt: 97…99 °C
Storage Temperature: Ambient
MDL Number: MFCD00672105
CAS Number: 73364-10-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR