1-n-Butyl-4-[(4-butylphenyl)ethynyl]benzene ≥99%
Supplier: Thermo Fisher Scientific
H27776.09
H27776.03
H27776.09EA
333
EUR
H27776.09
H27776.03
1-n-Butyl-4-[(4-butylphenyl)ethynyl]benzene ≥99%
1-n-Butyl-4-[(4-butylphenyl)ethynyl]benzene
Purity: 99+%
CAS No.: 80221-11-0
Molecular Formula: C22H26
Formula Weight: 290.45
Melting Point: 40-42°
Boiling Point: 172-174°/0.5mm
MDL No.: MFCD04038794
Formula:
C₂₂H₂₆ Boiling Pt: 172…174 °C (0,5 mmHg) Melting Pt: 40…42 °C Storage Temperature: Ambient |
MDL Number:
MFCD04038794 CAS Number: 80221-11-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...