3,4-Dihydro-2H-1,5-benzodioxepin-7-amine 98%

Supplier: Thermo Scientific
Danger

Synonyms: 7-Amino-3,4-dihydro-2H-1,5-benzodioxepine

H27862.06 H27862.03
H27862.06EA 93 EUR
H27862.06 H27862.03
3,4-Dihydro-2H-1,5-benzodioxepin-7-amine 98%
3,4-Dihydro-2H-1,5-benzodioxepin-7-amine

Purity: 98% CAS No.: 175136-34-2 Molecular Formula: C9H11NO2 Formula Weight: 165.19 Flash Point: 110°(230°F) Melting Point: 79-81° Boiling Point: 123-128°/0.2mm MDL No.: MFCD00218806

Formula: C₉H₁₁NO₂
MW: 165,19 g/mol
Boiling Pt: 123…128 °C (0,2 mmHg)
Melting Pt: 79…81 °C
Flash Pt: >110 °C (230 °F)
Storage Temperature: Ambient
MDL Number: MFCD00218806
CAS Number: 175136-34-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR