5-Aminopyridin-2(1H)-one 97%

Supplier: Thermo Scientific
Warning

Synonyms: 5-Amino-2-hydroxypyridine, 3-Amino-6-hydroxypyridine, 5-Amino-2(1H)-pyridone

H27961.06 H27961.03
H27961.06EA 292 EUR
H27961.06 H27961.03
5-Aminopyridin-2(1H)-one 97%
5-Aminopyridin-2(1H)-one

Purity: 97% CAS No.: 33630-94-3 Molecular Formula: C5H6N2O Formula Weight: 110.12 Storage Temperature: 2°C to 8°C Melting Point: ca 180° dec. MDL No.: MFCD03427652

Formula: C₅H₆N₂O
MW: 110,12 g/mol
Melting Pt: ∼ 180 °C (dec.)
MDL Number: MFCD03427652
CAS Number: 33630-94-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR