3-Hydroxypyrazine-2-carboxamide 98%

Supplier: Thermo Scientific
Warning

Synonyms: T-1105

H28250.06 H28250.03
H28250.06EA 153 EUR
H28250.06 H28250.03
3-Hydroxypyrazine-2-carboxamide 98%
3-Hydroxypyrazine-2-carboxamide

Purity: 98% CAS No.: 55321-99-8 Molecular Formula: C5H5N3O2 Formula Weight: 139.12 Melting Point: ca 270° dec. MDL No.: MFCD00233977

Formula: C₅H₅N₃O₂
MW: 139,114 g/mol
Melting Pt: ∼ 270 °C (dec.)
Storage Temperature: Ambient
MDL Number: MFCD00233977
CAS Number: 55321-99-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR