1-(p-Toluenesulphonyl)pyrrole 98%

Supplier: Thermo Scientific
Warning

Synonyms: 1-Tosylpyrrole, 1-Tosyl-1H-pyrrole, N-Tosylpyrrole

H28641.14 H28641.06
H28641.14EA 164 EUR
H28641.14 H28641.06
1-(p-Toluenesulphonyl)pyrrole 98%
1-(p-Toluenesulphonyl)pyrrole

Purity: 98% CAS No.: 17639-64-4 Molecular Formula: C11H11NO2S Formula Weight: 221.28 Melting Point: 99-103° MDL No.: MFCD00145014

Formula: C₁₁H₁₁NO₂S
MW: 221,27 g/mol
Melting Pt: 99…103 °C
Storage Temperature: Ambient
MDL Number: MFCD00145014
CAS Number: 17639-64-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR