tert-Butyl-3-(aminomethyl)pyrrolidine-1-carboxylate 95%
Supplier: Thermo Scientific
|
Warning
|
Synonyms:
3-(Aminomethyl)pyrrolidine, N1-BOC protected, tert-Butyl 3-(aminomethyl)pyrrolidine-1-carboxylate
H28674.MD
H28674.06
H28674.03
H28674.MDEA
73.5
EUR
H28674.MD
H28674.06
H28674.03
tert-Butyl-3-(aminomethyl)pyrrolidine-1-carboxylate 95%
tert-Butyl-3-(aminomethyl)pyrrolidine-1-carboxylate
Purity: 95%
CAS No.: 270912-72-6
Molecular Formula: C10H20N2O2
Formula Weight: 200.28
Refractive Index: 1.4740
MDL No.: MFCD01861798
Formula:
C₁₀H₂₀N₂O₂ MW: 200,28 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD01861798 CAS Number: 270912-72-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...