N-Boc-azetidine-3-carboxylic acid 97%

Supplier: Thermo Scientific
Warning

Synonyms: Boc-azetidine-3-carboxylic acid, N-Boc-Azetidine-3-carboxylic acid, 1-N-Boc-3-Azetidinecarboxylic acid, 1-Boc-azetidine-3-carboxylic Acid

H28817.06 H28817.03
H28817.06EA 862 EUR
H28817.06 H28817.03
N-Boc-azetidine-3-carboxylic acid 97%
N-Boc-azetidine-3-carboxylic acid

Purity: 97% CAS No.: 142253-55-2 Molecular Formula: C9H15NO4 Formula Weight: 201.22 Melting Point: 98-105° MDL No.: MFCD01860897

Formula: C₉H₁₅NO₄
MW: 201,22 g/mol
Melting Pt: 98…105 °C
Storage Temperature: Ambient
MDL Number: MFCD01860897
CAS Number: 142253-55-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR