tert-Butyl vinyl ether 98% stabilised
Supplier: Thermo Scientific
|
Danger
|
Stabilisation:
Stabilised with N,N-diethylaniline ∼ 0,1%
H30770.06
H30770.14
H30770.06EA
70
EUR
H30770.06
H30770.14
tert-Butyl vinyl ether 98% stabilised
tert-Butyl vinyl ether
Stab. with ca 0.1% N,N-diethylaniline
Purity: 98%
CAS No.: 926-02-3
Molecular Formula: C6H12O
Formula Weight: 100.16
Storage Temperature: 2°C to 8°C
Flash Point: -17°(1°F)
Refractive Index: 1.3990
Odour: Characteristic
Boiling Point: 75-76°
UN No.: UN3271
MDL No.: MFCD00048246
Formula:
C₆H₁₂O MW: 100,161 g/mol Boiling Pt: 75…76 °C Density: 0,762 Flash Pt: –17 °C (1 °F) Storage Temperature: Refrigerator |
MDL Number:
MFCD00048246 CAS Number: 926-02-3 EINECS: 618-860-6 UN: 3271 ADR: 3,II |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...