1-(4-Fluoro-3-methoxyphenyl)ethanol 95%

Supplier: Thermo Scientific

H32123.03 H32123.06
H32123.03EA 171 EUR
H32123.03 H32123.06
1-(4-Fluoro-3-methoxyphenyl)ethanol 95%
1-(4-Fluoro-3-methoxyphenyl)ethanol

Purity: 95% CAS No.: 870849-56-2 Molecular Formula: C9H11FO2 Formula Weight: 170.18 Storage Temperature: 2°C to 8°C MDL No.: MFCD08062372

Formula: C₉H₁₁FO₂
MW: 170,18 g/mol
MDL Number: MFCD08062372
CAS Number: 870849-56-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR