N-(4'-Methoxy-4-biphenylylsulphonyl)glycine 96%

Supplier: Thermo Scientific
Warning

H32299.03
H32299.03EA 166 EUR
H32299.03
N-(4'-Methoxy-4-biphenylylsulphonyl)glycine 96%
N-(4'-Methoxy-4-biphenylylsulphonyl)glycine

Purity: 96% CAS No.: 885269-46-5 Molecular Formula: C15H15NO5S Formula Weight: 321.35 Melting Point: 185-186° MDL No.: MFCD06409460

Formula: C₁₅H₁₅NO₅S
MW: 321,35 g/mol
Melting Pt: 185…186 °C
Storage Temperature: Ambient
MDL Number: MFCD06409460
CAS Number: 885269-46-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR