2-Nitro-4-pyridinecarboxylic acid 97%
Supplier: Thermo Fisher Scientific
|
Warning
|
H32361.MD
H32361.03
H32361.MDEA
0
EUR
H32361.MD
H32361.03
2-Nitro-4-pyridinecarboxylic acid 97%
2-Nitro-4-pyridinecarboxylic acid
Purity: 97%
CAS No.: 33225-74-0
Molecular Formula: C6H4N2O4
Formula Weight: 168.11
Melting Point: ca 170° dec.
MDL No.: MFCD04114160
Formula:
C₆H₄N₂O₄ Melting Pt: ∼ 170 °C (dec.) Storage Temperature: Ambient |
MDL Number:
MFCD04114160 CAS Number: 33225-74-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...