N-(4'-Methoxy-4-biphenylylsulphonyl)-L-valine 96%
Supplier: Thermo Scientific
|
Warning
|
H32521.03
H32521.03EA
166
EUR
H32521.03
N-(4'-Methoxy-4-biphenylylsulphonyl)-L-valine 96%
N-(4'-Methoxy-4-biphenylylsulphonyl)-L-valine
Purity: 96%
Molecular Formula: C18H21NO5S
Formula Weight: 363.44
Melting Point: 177-179°
MDL No.: MFCD06409462
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...